Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022785.3 | NP_073622.2 | 8 | 1054 | ||
NM_198856.2 | NP_942153.1 | 6 | 696 | |||
XM_005261704.2 | XP_005261761.1 | 8 | 994 | |||
XM_005261705.3 | XP_005261762.1 | 7 | 908 | |||
XM_011530316.1 | XP_011528618.1 | 9 | 1281 | |||
XM_011530317.2 | XP_011528619.1 | 7 | 841 | |||
XM_011530318.2 | XP_011528620.1 | 7 | 937 | |||
XM_011530319.2 | XP_011528621.1 | 6 | 629 | |||
XM_011530320.2 | XP_011528622.1 | 9 | 1093 | |||
XM_011530321.2 | XP_011528623.1 | 6 | 624 | |||
XM_011530322.2 | XP_011528624.1 | 7 | 752 | |||
XM_011530323.2 | XP_011528625.1 | 4 | 410 | |||
XM_011530325.2 | XP_011528627.1 | 2 | 155 | |||
XM_011530326.2 | XP_011528628.1 | 9 | 1093 | |||
XM_011530328.2 | XP_011528630.1 | 9 | 1093 | |||
XM_011530330.2 | XP_011528632.1 | 9 | 1093 | |||
XM_017028910.1 | XP_016884399.1 | 9 | 1093 | |||
XM_017028911.1 | XP_016884400.1 | 9 | 1093 | |||
XM_017028912.1 | XP_016884401.1 | 5 | 752 | |||
GenBank mRNA | AK058069.1 | - | 8 | 1054 | ||
AK292411.1 | - | 8 | 994 | |||
AK292498.1 | - | 6 | 673 | |||
AK296679.1 | - | 6 | 605 | |||
AK311267.1 | - | 1 | 65 | |||
BC039315.1 | - | 6 | 549 | 5297436 | ||
CD106996.1 | - | 2 | 216 | |||
Y10776.1 | - | 5 | 498 |