Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022786.1 | NP_073623.1 | 2 | 262 | ||
XM_017002106.1 | XP_016857595.1 | 2 | 257 | |||
GenBank mRNA | AF271780.1 | - | 2 | 262 | ||
AF290878.1 | - | 2 | 262 | |||
AF321442.1 | - | 2 | 296 | |||
AK026629.1 | - | 2 | 250 | |||
AK293049.1 | - | 2 | 268 | |||
BC016309.1 | - | 2 | 257 | 4047121 |