Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022789.3 | NP_073626.1 | 2 | 1287 | ||
NM_172314.1 | NP_758525.1 | 3 | 1139 | |||
GenBank mRNA | AF305200.1 | - | 2 | 1287 | ||
AY359127.1 | - | 2 | 1287 |