Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134774.1 | NP_001128246.1 | 10 | 1268 | ||
NM_001134775.1 | NP_001128247.1 | 11 | 1562 | |||
NM_001134776.1 | NP_001128248.1 | 11 | 1441 | |||
NM_001318734.1 | NP_001305663.1 | 11 | 1499 | |||
NM_022822.2 | NP_073733.1 | 11 | 1536 | |||
XM_005274183.1 | XP_005274240.1 | 11 | 1478 | |||
XM_005274184.1 | XP_005274241.1 | 11 | 1499 | |||
XM_017018133.1 | XP_016873622.1 | 9 | 1435 | |||
GenBank mRNA | AK022449.1 | - | 11 | 1428 | ||
AK022907.1 | - | 11 | 1481 | |||
AK094593.1 | - | 9 | 1138 | |||
AK126489.1 | - | 9 | 1920 | |||
AK315725.1 | - | 11 | 1431 | |||
AL136864.1 | - | 11 | 1462 | |||
BC034373.1 | - | 11 | 1424 | 4342879 |