Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282805.1 | NP_001269734.1 | 7 | 1778 | ||
NM_001282806.1 | NP_001269735.1 | 4 | 1482 | |||
NM_001282807.1 | NP_001269736.1 | 4 | 1525 | |||
NM_022826.3 | NP_073737.1 | 5 | 1650 | |||
XM_005246773.2 | XP_005246830.1 | 5 | 1673 | |||
XM_005246774.1 | XP_005246831.1 | 5 | 1673 | |||
XM_017004718.1 | XP_016860207.1 | 5 | 1650 | |||
XM_017004719.1 | XP_016860208.1 | 4 | 1508 | |||
XM_017004720.1 | XP_016860209.1 | 4 | 1470 | |||
XM_017004721.1 | XP_016860210.1 | 4 | 1500 | |||
GenBank mRNA | AK022973.1 | - | 5 | 1608 | ||
AK222880.1 | - | 5 | 1600 | |||
AK292836.1 | - | 7 | 1746 | |||
AK299051.1 | - | 4 | 1450 | |||
AK302347.1 | - | 4 | 1474 | |||
AK316382.1 | - | 4 | 1482 | |||
AL597638.1 | - | 1 | 263 | |||
AL833651.1 | - | 1 | 651 | |||
BC003404.1 | - | 1 | 49 | |||
BC065014.1 | - | 5 | 1574 | 6169288 |