Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011554.2 | NP_001011554.1 | 8 | 1010 | ||
NM_001193339.1 | NP_001180268.1 | 6 | 824 | |||
NM_001193340.1 | NP_001180269.1 | 8 | 1010 | |||
NM_001193342.1 | NP_001180271.1 | 7 | 801 | |||
NM_022829.5 | NP_073740.2 | 7 | 974 | |||
GenBank mRNA | AF154121.1 | - | 7 | 953 | ||
AK056713.1 | - | 7 | 972 | |||
AK096658.1 | - | 8 | 1010 | |||
AK293902.1 | - | 7 | 790 | |||
AK295748.1 | - | 6 | 822 | |||
AK310960.1 | - | 6 | 984 | |||
AK315131.1 | - | 7 | 972 | |||
AY072810.3 | - | 7 | 953 | |||
BC035966.1 | - | 7 | 972 | 4609776 | ||
BM474324.1 | - | 405 |