Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022834.4 | NP_073745.2 | 3 | 2506 | ||
NM_199121.2 | NP_954572.2 | 3 | 2111 | |||
GenBank mRNA | AK025868.1 | - | 1 | 1652 | ||
AK075366.1 | - | 3 | 2330 | |||
AK096773.1 | - | 3 | 1918 | |||
AL137722.1 | - | 1 | 656 | |||
BC003543.1 | - | 2 | 1790 | |||
BC059409.1 | - | 3 | 2313 | 30349211 |