Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168478.1 | NP_001161950.1 | 4 | 859 | ||
NM_001168479.1 | NP_001161951.1 | 6 | 975 | |||
NM_001168480.1 | NP_001161952.1 | 6 | 969 | |||
NM_001168482.1 | NP_001161954.1 | 3 | 758 | |||
NM_001168485.1 | NP_001161957.1 | 5 | 856 | |||
NM_022838.3 | NP_073749.2 | 3 | 746 | |||
GenBank mRNA | AK023031.1 | - | 2 | 723 | ||
AK096692.1 | - | 6 | 969 | |||
AL537480.3 | - | 4 | 779 | |||
BC058904.1 | - | 3 | 701 | 5531406 | ||
BC064983.1 | - | 3 | 1014 | 2960871 | ||
BX352705.2 | - | 3 | 734 | |||
CR749476.1 | - | 1 | 2711 | |||
DA684921.1 | - | 2 | 482 |