Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022842.4 | NP_073753.3 | 4 | 1122 | ||
NM_178181.2 | NP_835488.1 | 4 | 1122 | |||
XM_011534024.1 | XP_011532326.1 | 4 | 1080 | |||
XM_017007070.1 | XP_016862559.1 | 4 | 1108 | |||
GenBank mRNA | AF468010.1 | - | 4 | 1097 | ||
AK023834.1 | - | 2 | 700 | |||
AK026187.1 | - | 1 | 139 | |||
AK026622.1 | - | 4 | 1097 | |||
AK314441.1 | - | 4 | 1086 | |||
AY026461.1 | - | 4 | 1069 | |||
AY167484.1 | - | 4 | 1079 | |||
AY358779.1 | - | 4 | 1080 | |||
AY375452.1 | - | 4 | 1068 | |||
BC021099.1 | - | 4 | 1122 | |||
BC069254.1 | - | 4 | 1075 | |||
KU215672.1 | - | 3 | 900 | |||
KU257676.1 | - | 3 | 897 |