Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301860.1 | NP_001288789.1 | 10 | 1599 | ||
NR_126051.1 | 10 | 1599 | ||||
XM_005260516.2 | XP_005260573.1 | 10 | 1565 | |||
XM_006723863.2 | XP_006723926.1 | 11 | 1682 | |||
XM_011528996.1 | XP_011527298.1 | 10 | 1597 | |||
XM_011528997.2 | XP_011527299.1 | 11 | 1803 | |||
XM_011528998.2 | XP_011527300.1 | 10 | 1620 | |||
XM_011528999.2 | XP_011527301.1 | 11 | 1750 | |||
XM_011529000.1 | XP_011527302.1 | 10 | 1594 | |||
XM_011529001.1 | XP_011527303.1 | 10 | 1597 | |||
XM_011529002.2 | XP_011527304.1 | 9 | 1399 | |||
XM_011529003.1 | XP_011527305.1 | 10 | 1597 | |||
XM_011529004.1 | XP_011527306.1 | 10 | 1597 | |||
XM_011529005.2 | XP_011527307.1 | 10 | 1597 | |||
XM_017028020.1 | XP_016883509.1 | 10 | 1594 | |||
GenBank mRNA | BC140806.1 | - | 10 | 1480 | 9021674 | |
BX647082.1 | - | 9 | 1941 |