Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022897.4 | NP_075048.1 | 18 | 2178 | ||
XM_011534627.1 | XP_011532929.1 | 18 | 2034 | |||
XM_011534631.2 | XP_011532933.1 | 17 | 2047 | |||
XM_017009736.1 | XP_016865225.1 | 17 | 1914 | |||
XM_017009737.1 | XP_016865226.1 | 17 | 2047 | |||
XM_017009738.1 | XP_016865227.1 | 17 | 1887 | |||
XM_017009739.1 | XP_016865228.1 | 17 | 1887 | |||
XM_017009740.1 | XP_016865229.1 | 16 | 1767 | |||
XM_017009741.1 | XP_016865230.1 | 18 | 2034 | |||
XM_017009742.1 | XP_016865231.1 | 18 | 2034 | |||
XM_017009743.1 | XP_016865232.1 | 18 | 2208 | |||
XM_017009744.1 | XP_016865233.1 | 18 | 2034 | |||
XM_017009745.1 | XP_016865234.1 | 10 | 1231 | |||
XM_017009747.1 | XP_016865236.1 | 6 | 506 | |||
GenBank mRNA | AF222747.1 | - | 18 | 2034 | ||
AJ271459.1 | - | 18 | 2042 | |||
AJ288952.1 | - | 21 | 2381 | |||
AJ288953.1 | - | 20 | 2220 | |||
AJ288954.1 | - | 20 | 2341 | |||
AJ288955.1 | - | 19 | 2160 | |||
AK027880.1 | - | 16 | 1862 | |||
AK057478.1 | - | 3 | 373 | |||
AK315159.1 | - | 19 | 2246 | |||
AL832980.1 | - | 18 | 2077 |