Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022903.3 | NP_075054.3 | 2 | 941 | ||
GenBank mRNA | AK022862.1 | - | 2 | 912 | ||
AK023691.1 | - | 1 | 196 | |||
AK098658.1 | - | 3 | 776 | |||
BC035516.1 | - | 2 | 910 | 5229681 | ||
BC071960.1 | - | 1 | 100 |