Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022904.1 | NP_075055.1 | 17 | 2936 | ||
XM_011528185.1 | XP_011526487.1 | 17 | 2945 | |||
XM_011528186.1 | XP_011526488.1 | 17 | 2927 | |||
GenBank mRNA | AK024488.1 | - | 15 | 3933 | ||
AK025091.1 | - | 4 | 1258 | |||
AK122587.1 | - | 17 | 2931 | |||
AL832868.1 | - | 3 | 393 | |||
BC030281.1 | - | 4 | 686 |