Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030811.3 | NP_110438.1 | 4 | 967 | ||
GenBank mRNA | AF308301.1 | - | 4 | 918 | ||
BC013018.2 | - | 4 | 934 | 3051126 |