Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321970.1 | NP_001308899.1 | 4 | 563 | ||
NM_001321972.1 | NP_001308901.1 | 4 | 1687 | |||
NM_022839.4 | NP_073750.2 | 4 | 566 | |||
NM_176805.3 | NP_789775.1 | 3 | 467 | |||
NR_135915.1 | 3 | 547 | ||||
NR_135917.1 | 4 | 546 | ||||
GenBank mRNA | AB049944.1 | - | 4 | 566 | ||
AK026165.1 | - | 4 | 311 | |||
AK027059.1 | - | 4 | 1432 | |||
AK222933.1 | - | 4 | 311 | |||
AK223152.1 | - | 4 | 311 | |||
AK315407.1 | - | 4 | 311 | |||
BC009451.1 | - | 3 | 199 | |||
BC012489.1 | - | 4 | 320 | 4452949 | ||
BC032378.1 | - | 4 | 335 | 5110633 | ||
DA803430.1 | - | 3 | 295 |