Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032478.3 | NP_115867.2 | 2, 3 | 475 | ||
GenBank mRNA | AK313283.1 | - | 2, 3 | 290 | ||
BC013311.2 | - | 2, 3 | 274 | 3546083 | ||
BC034460.1 | - | 2, 3 | 229 | |||
DB289191.1 | - | 2, 3 | 475 |