Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG4 SIRNA Assays ›
Gene Symbol:
NDRG4
Gene Name:
NDRG family member 4
Gene Aliases:
BDM1, SMAP-8, SMAP8
Chromosome Location:
Chr.16: 58463645 - 58515403 on Build GRCh38
UniGene ID:
Hs.322430
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35159
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130487.1
NP_001123959.1
6, 7
808
NM_001242833.1
NP_001229762.1
5, 6
507
NM_001242834.1
NP_001229763.1
4, 5
471
NM_001242835.1
NP_001229764.1
4, 5
417
NM_001242836.1
NP_001229765.1
4, 5
417
NM_020465.3
NP_065198.1
6, 7
583
NM_022910.3
NP_075061.1
6, 7
680
NR_040072.1
4, 5
509
XM_006721252.3
XP_006721315.1
7, 8
683
XM_006721253.3
XP_006721316.1
7, 8
616
XM_006721254.3
XP_006721317.1
6, 7
670
XM_006721256.3
XP_006721319.1
7, 8
614
XM_006721257.3
XP_006721320.1
6, 7
655
XM_006721258.3
XP_006721321.1
6, 7
559
XM_006721259.3
XP_006721322.1
6, 7
624
XM_006721260.3
XP_006721323.1
6, 7
562
XM_006721261.3
XP_006721324.1
5, 6
507
XM_006721262.3
XP_006721325.1
4, 5
588
XM_011523290.2
XP_011521592.1
7, 8
770
XM_011523291.2
XP_011521593.1
6, 7
734
XM_011523292.2
XP_011521594.1
7, 8
770
XM_011523293.2
XP_011521595.1
6, 7
728
XM_011523294.2
XP_011521596.1
7, 8
549
XM_017023582.1
XP_016879071.1
6, 7
734
XM_017023583.1
XP_016879072.1
7, 8
770
XM_017023584.1
XP_016879073.1
7, 8
683
XM_017023585.1
XP_016879074.1
6, 7
670
XM_017023586.1
XP_016879075.1
6, 7
617
XM_017023587.1
XP_016879076.1
7, 8
614
XM_017023588.1
XP_016879077.1
6, 7
559
XM_017023589.1
XP_016879078.1
6, 7
624
XM_017023590.1
XP_016879079.1
7, 8
549
GenBank mRNA
AB021172.1
-
6, 7
483
AB033006.2
-
4, 5
403
AB044944.1
-
4, 5
417
AB044945.1
-
4, 5
417
AB044946.1
-
5, 6
430
AF308608.1
-
4, 5
424
AK055038.1
-
4, 5
419
AK126574.1
-
6, 7
580
AK126729.1
-
7, 8
2530
AK131248.1
-
4, 5
471
AK296415.1
-
4, 5
471
AK299949.1
-
5, 6
507
AK316089.1
-
4, 5
509
AL136584.1
-
4, 5
403
BC001527.1
-
4, 5
401
BC011795.2
-
4, 5
405
4139594
CF552009.1
-
6, 7
526
CR936733.1
-
6, 7
494
KU178760.1
-
4, 5
311
KU178761.1
-
4, 5
311
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
heart looping
response to stress
brain development
visual learning
negative regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of neuron projection development
negative regulation of smooth muscle cell migration
cell growth
cell differentiation
embryonic heart tube development
vesicle docking
negative regulation of smooth muscle cell proliferation
cardiac muscle cell proliferation
cell migration involved in heart development
positive regulation of ERK1 and ERK2 cascade
regulation of endocytic recycling
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline