Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040454.1 | NP_001035544.1 | 13 | 1758 | ||
NM_001281732.1 | NP_001268661.1 | 13 | 1562 | |||
NM_001281733.1 | NP_001268662.1 | 12 | 1349 | |||
NM_022911.2 | NP_075062.2 | 14 | 1670 | |||
NM_134263.2 | NP_599025.2 | 14 | 1670 | |||
NM_134426.2 | NP_602298.2 | 14 | 1670 | |||
GenBank mRNA | AB033288.1 | - | 14 | 1633 | ||
AB102713.1 | - | 14 | 1715 | |||
AB208936.1 | - | 8 | 3519 | |||
AF279265.1 | - | 13 | 1758 | |||
AF288410.1 | - | 14 | 1618 | |||
AF416721.1 | - | 14 | 1618 | |||
AK056237.1 | - | 12 | 1349 | |||
AK297695.1 | - | 13 | 1510 | |||
BC017697.1 | - | 14 | 1619 | 4398446 |