Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040454.1 | NP_001035544.1 | 17, 18 | 2262 | ||
NM_001281732.1 | NP_001268661.1 | 17, 18 | 2066 | |||
NM_001281733.1 | NP_001268662.1 | 16, 17 | 1850 | |||
NM_022911.2 | NP_075062.2 | 18, 19 | 2174 | |||
NM_134263.2 | NP_599025.2 | 18, 19 | 2171 | |||
NM_134426.2 | NP_602298.2 | 17, 18 | 2117 | |||
GenBank mRNA | AB033288.1 | - | 17, 18 | 2080 | ||
AB102713.1 | - | 17, 18 | 2162 | |||
AB208936.1 | - | 9, 10 | 5215 | |||
AF279265.1 | - | 17, 18 | 2262 | |||
AF288410.1 | - | 18, 19 | 2122 | |||
AF416721.1 | - | 18, 19 | 2119 | |||
AK056237.1 | - | 16, 17 | 1850 | |||
AK297695.1 | - | 17, 18 | 2014 | |||
BC017697.1 | - | 18, 19 | 2120 | 4398446 |