Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 35 | 5945 | ||
XM_005246787.3 | XP_005246844.1 | 35 | 5971 | |||
XM_005246788.2 | XP_005246845.1 | 14 | 2447 | |||
XM_006712698.3 | XP_006712761.1 | 36 | 6058 | |||
XM_006712699.3 | XP_006712762.1 | 36 | 6058 | |||
XM_011511658.2 | XP_011509960.1 | 36 | 6013 | |||
XM_011511659.2 | XP_011509961.1 | 36 | 6070 | |||
XM_011511660.2 | XP_011509962.1 | 32 | 5392 | |||
XM_011511662.2 | XP_011509964.1 | 28 | 4727 | |||
XM_011511663.2 | XP_011509965.1 | 36 | 6048 | |||
XM_017004728.1 | XP_016860217.1 | 23 | 4125 | |||
XM_017004729.1 | XP_016860218.1 | 11 | 2139 | |||
GenBank mRNA | AK127772.1 | - | 9 | 1813 | ||
AK128636.1 | - | 5 | 672 | |||
AY172970.1 | - | 5 | 851 |