Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 41 | 6623 | ||
XM_005246787.3 | XP_005246844.1 | 41 | 6649 | |||
XM_005246788.2 | XP_005246845.1 | 20 | 3125 | |||
XM_006712698.3 | XP_006712761.1 | 42 | 6736 | |||
XM_006712699.3 | XP_006712762.1 | 42 | 6736 | |||
XM_011511658.2 | XP_011509960.1 | 42 | 6691 | |||
XM_011511659.2 | XP_011509961.1 | 42 | 6748 | |||
XM_011511660.2 | XP_011509962.1 | 38 | 6070 | |||
XM_011511662.2 | XP_011509964.1 | 34 | 5405 | |||
XM_011511663.2 | XP_011509965.1 | 42 | 6726 | |||
XM_017004728.1 | XP_016860217.1 | 29 | 4803 | |||
XM_017004729.1 | XP_016860218.1 | 17 | 2817 | |||
GenBank mRNA | AK127772.1 | - | 15 | 2491 | ||
AK128636.1 | - | 11 | 1350 | |||
AY172970.1 | - | 11 | 1529 | |||
BX647133.1 | - | 5 | 450 |