Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023004.5 | NP_075380.1 | 2 | 826 | ||
GenBank mRNA | AF283463.1 | - | 2 | 637 | ||
AK054602.1 | - | 2 | 882 | |||
AL834449.1 | - | 2 | 769 | |||
BC011787.1 | - | 2 | 671 | 4040540 | ||
CR456360.1 | - | 2 | 665 |