Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023014.1 | NP_075390.1 | 3 | 640 | ||
GenBank mRNA | AL049674.1 | - | 3 | 640 | ||
BC075008.2 | - | 2 | 589 | 30915615 | ||
BC075009.2 | - | 2 | 589 | 30915402 |
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023013.4 | NP_075389.2 | 3 | 656 | ||
GenBank mRNA | AK125568.1 | - | 3 | 656 | ||
AL049686.1 | - | 3 | 656 | |||
BC144199.1 | - | 3 | 626 | 9052719 |
Gene Symbol: PRAMEF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291380.1 | NP_001278309.1 | 3 | 656 |
Gene Symbol: PRAMEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024661.2 | NP_001019832.2 | 3 | 656 |