Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023080.2 | NP_075568.1 | 5 | 680 | ||
GenBank mRNA | AK024642.1 | - | 5 | 678 | ||
BC010001.2 | - | 5 | 640 | 4135341 | ||
BC015181.1 | - | 4 | 441 | |||
BX648996.1 | - | 4 | 740 | |||
CR457330.1 | - | 4 | 626 |