Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK33 SIRNA Assays ›
Gene Symbol:
STK33
Gene Name:
serine/threonine kinase 33
Gene Aliases:
-
Chromosome Location:
Chr.11: 8335029 - 8594289 on Build GRCh38
UniGene ID:
Hs.501833
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35284
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289058.1
NP_001275987.1
9
969
NM_001289059.1
NP_001275988.1
6
676
NM_001289061.1
NP_001275990.1
8
1234
NM_030906.3
NP_112168.1
9
1383
XM_011520288.2
XP_011518590.1
9
2461
XM_011520290.2
XP_011518592.1
11
1380
XM_011520292.2
XP_011518594.1
9
1182
XM_011520293.2
XP_011518595.1
9
1211
XM_011520294.1
XP_011518596.1
8
1176
XM_011520295.2
XP_011518597.1
8
3272
XM_011520297.2
XP_011518599.1
8
949
XM_017018143.1
XP_016873632.1
11
1440
XM_017018144.1
XP_016873633.1
11
1342
XM_017018145.1
XP_016873634.1
10
3374
XM_017018146.1
XP_016873635.1
11
1504
XM_017018147.1
XP_016873636.1
10
3301
XM_017018148.1
XP_016873637.1
9
3233
XM_017018149.1
XP_016873638.1
10
1385
XM_017018150.1
XP_016873639.1
10
1240
XM_017018151.1
XP_016873640.1
10
1199
XM_017018152.1
XP_016873641.1
8
1530
XM_017018153.1
XP_016873642.1
8
1128
XM_017018154.1
XP_016873643.1
8
987
XM_017018155.1
XP_016873644.1
8
1542
XM_017018156.1
XP_016873645.1
8
988
XM_017018157.1
XP_016873646.1
8
753
XM_017018158.1
XP_016873647.1
9
1039
XM_017018159.1
XP_016873648.1
10
3377
XM_017018160.1
XP_016873649.1
8
1176
XM_017018161.1
XP_016873650.1
10
3383
XM_017018162.1
XP_016873651.1
9
986
XM_017018163.1
XP_016873652.1
8
977
XM_017018164.1
XP_016873653.1
8
863
XM_017018165.1
XP_016873654.1
9
942
GenBank mRNA
AJ303380.2
-
7
1727
AK092720.1
-
6
615
AK093251.1
-
8
1092
AK122808.1
-
5
3302
AK293193.1
-
9
835
AK310754.1
-
2
199
AL833011.1
-
2
73
BC031231.1
-
9
1331
5296502
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00294893_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein autophosphorylation
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline