Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK33 SIRNA Assays ›
Gene Symbol:
STK33
Gene Name:
serine/threonine kinase 33
Gene Aliases:
-
Chromosome Location:
Chr.11: 8335029 - 8594289 on Build GRCh38
UniGene ID:
Hs.501833
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35285
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289058.1
NP_001275987.1
12
1283
NM_001289059.1
NP_001275988.1
9
990
NM_001289061.1
NP_001275990.1
11
1548
NM_030906.3
NP_112168.1
12
1697
XM_011520288.2
XP_011518590.1
12
2775
XM_011520290.2
XP_011518592.1
14
1694
XM_011520292.2
XP_011518594.1
12
1496
XM_011520293.2
XP_011518595.1
12
1525
XM_011520294.1
XP_011518596.1
11
1490
XM_011520295.2
XP_011518597.1
11
3586
XM_011520297.2
XP_011518599.1
11
1263
XM_017018143.1
XP_016873632.1
14
1754
XM_017018144.1
XP_016873633.1
14
1656
XM_017018145.1
XP_016873634.1
13
3688
XM_017018146.1
XP_016873635.1
14
1818
XM_017018147.1
XP_016873636.1
13
3615
XM_017018148.1
XP_016873637.1
12
3547
XM_017018149.1
XP_016873638.1
13
1699
XM_017018150.1
XP_016873639.1
13
1554
XM_017018151.1
XP_016873640.1
13
1513
XM_017018152.1
XP_016873641.1
11
1844
XM_017018153.1
XP_016873642.1
11
1442
XM_017018154.1
XP_016873643.1
11
1301
XM_017018155.1
XP_016873644.1
11
1856
XM_017018156.1
XP_016873645.1
11
1302
XM_017018157.1
XP_016873646.1
11
1067
XM_017018158.1
XP_016873647.1
12
1353
XM_017018159.1
XP_016873648.1
13
3691
XM_017018160.1
XP_016873649.1
11
1490
XM_017018161.1
XP_016873650.1
13
3697
XM_017018162.1
XP_016873651.1
12
1300
XM_017018163.1
XP_016873652.1
11
1291
XM_017018164.1
XP_016873653.1
11
1177
XM_017018165.1
XP_016873654.1
12
1256
GenBank mRNA
AJ303380.2
-
10
2041
AK092720.1
-
9
929
AK093251.1
-
11
1406
AK122808.1
-
8
3616
AK293193.1
-
12
1149
AK310754.1
-
5
513
AL833011.1
-
5
387
BC031231.1
-
12
1645
5296502
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein autophosphorylation
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline