Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35298
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
8
2486
NM_001206856.2
NP_001193785.1
8
2486
NM_001319842.1
NP_001306771.1
7
2403
NM_001319843.1
NP_001306772.1
8
2486
NM_001319844.1
NP_001306773.1
9
1741
NM_001319845.1
NP_001306774.1
9
1741
NM_001319846.1
NP_001306775.1
9
1731
NM_023925.4
NP_076414.2
8
2486
NM_032156.4
NP_115532.3
8
2486
XM_006719138.3
XP_006719201.1
8
2609
XM_006719139.3
XP_006719202.1
8
2609
XM_006719140.3
XP_006719203.1
8
2609
XM_006719142.3
XP_006719205.1
8
2609
XM_006719144.3
XP_006719207.1
8
2609
XM_006719145.3
XP_006719208.1
8
2609
XM_006719146.3
XP_006719209.1
8
2609
XM_006719147.3
XP_006719210.1
8
2609
XM_017019851.1
XP_016875340.1
8
2609
XM_017019852.1
XP_016875341.1
9
1704
XM_017019853.1
XP_016875342.1
9
1774
XM_017019854.1
XP_016875343.1
8
2609
XM_017019855.1
XP_016875344.1
9
1731
XM_017019856.1
XP_016875345.1
9
1731
XM_017019857.1
XP_016875346.1
9
1704
XM_017019858.1
XP_016875347.1
9
1731
XM_017019859.1
XP_016875348.1
9
1704
XM_017019860.1
XP_016875349.1
8
2609
XM_017019861.1
XP_016875350.1
9
1731
XM_017019862.1
XP_016875351.1
8
2609
XM_017019863.1
XP_016875352.1
8
2609
XM_017019864.1
XP_016875353.1
8
2609
XM_017019865.1
XP_016875354.1
8
2609
XM_017019866.1
XP_016875355.1
9
1704
XM_017019867.1
XP_016875356.1
9
1731
XM_017019868.1
XP_016875357.1
9
1704
XM_017019869.1
XP_016875358.1
9
1731
XM_017019870.1
XP_016875359.1
9
1704
XM_017019871.1
XP_016875360.1
9
1775
XM_017019872.1
XP_016875361.1
9
1731
XM_017019873.1
XP_016875362.1
9
1731
XM_017019874.1
XP_016875363.1
8
1621
XM_017019875.1
XP_016875364.1
9
1731
XM_017019876.1
XP_016875365.1
6
1232
XM_017019877.1
XP_016875366.1
6
1232
XM_017019878.1
XP_016875367.1
6
1232
XM_017019879.1
XP_016875368.1
8
1621
XM_017019880.1
XP_016875369.1
6
1232
XM_017019881.1
XP_016875370.1
8
1621
XM_017019882.1
XP_016875371.1
8
1621
XM_017019883.1
XP_016875372.1
6
1232
GenBank mRNA
AB058776.1
-
781
AF326778.1
-
3
781
AK021453.1
-
7
1253
AL834212.1
-
5
973
AY074490.1
-
8
2486
AY074491.1
-
7
2403
BC117672.1
-
8
2283
40068103
BC117673.1
-
8
2283
40068104
BX537569.1
-
7
1550
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-ptlgl:80/100.66.77.47:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0