Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319839.1 | NP_001306768.1 | 4, 5 | 684 | ||
NM_001319840.1 | NP_001306769.1 | 4, 5 | 684 | |||
NM_023928.4 | NP_076417.2 | 4, 5 | 684 | |||
XM_011538692.2 | XP_011536994.1 | 2, 3 | 195 | |||
GenBank mRNA | AB054121.1 | - | 4, 5 | 573 | ||
AK022451.1 | - | 4, 5 | 614 | |||
AK022740.1 | - | 2, 3 | 224 | |||
AK024036.1 | - | 4, 5 | 613 | |||
AK304176.1 | - | 5, 6 | 886 | |||
AK308779.1 | - | 5, 6 | 892 | |||
AL833047.1 | - | 3, 4 | 325 | |||
AY091468.1 | - | 4, 5 | 445 | |||
BC040490.1 | - | 4, 5 | 672 | 5295926 | ||
BC041000.1 | - | 4, 5 | 667 | 5273267 | ||
BC051862.1 | - | 4, 5 | 590 | 5762683 |