Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136469.2 | NP_001129941.1 | 5 | 1830 | ||
NM_001282865.1 | NP_001269794.1 | 3 | 486 | |||
NM_023943.3 | NP_076432.1 | 5 | 1834 | |||
XM_011513097.2 | XP_011511399.1 | 4 | 3836 | |||
XM_011513098.2 | XP_011511400.1 | 6 | 1929 | |||
XM_017007080.1 | XP_016862569.1 | 6 | 1986 | |||
XM_017007081.1 | XP_016862570.1 | 4 | 361 | |||
XM_017007082.1 | XP_016862571.1 | 3 | 2425 | |||
GenBank mRNA | AK095050.1 | - | 3 | 480 | ||
AL136578.1 | - | 2 | 1186 | |||
AY358381.1 | - | 5 | 1716 | |||
BC000568.2 | - | 5 | 1713 | 3163990 | ||
DB522014.1 | - | 1 | 44 |