Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023944.3 | NP_076433.3 | 5 | 617 | ||
NR_117085.1 | 4 | 635 | ||||
XM_006722850.3 | XP_006722913.1 | 5 | 502 | |||
XM_011528202.2 | XP_011526504.1 | 3 | 137 | |||
XM_011528203.2 | XP_011526505.1 | 2 | 156 | |||
XM_011528204.2 | XP_011526506.1 | 2 | 156 | |||
XM_011528205.2 | XP_011526507.1 | 1 | 12 | |||
XM_011528207.2 | XP_011526509.1 | 2 | 156 | |||
XM_011528208.2 | XP_011526510.1 | 4 | 256 | |||
XM_017027172.1 | XP_016882661.1 | 5 | 559 | |||
GenBank mRNA | AB035130.1 | - | 5 | 528 | ||
AB035131.1 | - | 5 | 528 | |||
AK075435.1 | - | 4 | 563 | |||
AK304138.1 | - | 5 | 560 | |||
AK304389.1 | - | 4 | 653 | |||
AL832171.1 | - | 5 | 470 | |||
AY008841.1 | - | 4 | 498 | |||
AY358977.1 | - | 5 | 539 | |||
BC035350.1 | - | 5 | 544 | 5187264 | ||
DB016100.1 | - | 5 | 507 |