Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023944.3 | NP_076433.3 | 8 | 1056 | ||
NR_117085.1 | 7 | 1074 | ||||
XM_006722850.3 | XP_006722913.1 | 8 | 941 | |||
XM_011528202.2 | XP_011526504.1 | 6 | 576 | |||
XM_011528203.2 | XP_011526505.1 | 5 | 1798 | |||
XM_011528204.2 | XP_011526506.1 | 6 | 1277 | |||
XM_011528205.2 | XP_011526507.1 | 5 | 489 | |||
XM_011528207.2 | XP_011526509.1 | 5 | 2185 | |||
XM_011528208.2 | XP_011526510.1 | 7 | 695 | |||
XM_017027172.1 | XP_016882661.1 | 8 | 998 | |||
GenBank mRNA | AB035130.1 | - | 8 | 967 | ||
AB035131.1 | - | 8 | 967 | |||
AK074412.1 | - | 2 | 2358 | |||
AK075435.1 | - | 7 | 1002 | |||
AL832171.1 | - | 7 | 1963 | |||
AY008841.1 | - | 7 | 937 | |||
AY358977.1 | - | 8 | 978 | |||
BC035350.1 | - | 8 | 983 | 5187264 |