Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142674.1 | NP_001136146.1 | 13 | 1318 | ||
NM_001142675.1 | NP_001136147.1 | 12 | 1428 | |||
NM_001142676.1 | NP_001136148.1 | 13 | 1289 | |||
NM_001142677.1 | NP_001136149.1 | 11 | 1121 | |||
NM_023947.3 | NP_076436.3 | 12 | 1214 | |||
XM_005253072.2 | XP_005253129.1 | 12 | 1301 | |||
XM_005253073.4 | XP_005253130.1 | 12 | 1209 | |||
XM_006718282.2 | XP_006718345.1 | 13 | 1289 | |||
XM_011520299.1 | XP_011518601.1 | 12 | 1525 | |||
XM_011520300.1 | XP_011518602.1 | 11 | 1432 | |||
XM_017018166.1 | XP_016873655.1 | 13 | 1375 | |||
GenBank mRNA | AF212229.1 | - | 13 | 1252 | ||
AK027711.1 | - | 12 | 1163 | |||
AK075413.1 | - | 13 | 1225 | |||
AK124697.1 | - | 12 | 1790 | |||
AK225025.1 | - | 13 | 1254 | |||
AK225398.1 | - | 13 | 1237 | |||
AK297743.1 | - | 12 | 1295 | |||
AK301245.1 | - | 12 | 1274 | |||
AY037151.1 | - | 11 | 1082 | |||
BC000001.2 | - | 13 | 1238 | 3504261 | ||
BC013642.1 | - | 10 | 850 | |||
BC095409.1 | - | 12 | 1150 | 30333427 |