Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321439.1 | NP_001308368.1 | 4 | 428 | ||
NM_001321440.1 | NP_001308369.1 | 4 | 428 | |||
NM_024029.4 | NP_076934.1 | 4 | 699 | |||
XM_011528270.2 | XP_011526572.1 | 4 | 428 | |||
GenBank mRNA | BC000056.2 | - | 4 | 347 | 3506385 | |
BC013014.2 | - | 4 | 368 | 2960233 | ||
BM554546.1 | - | 4 | 346 | |||
HY020827.1 | - | 4 | 433 |