Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243749.1 | NP_001230678.1 | 4 | 1425 | ||
NM_001243751.1 | NP_001230680.1 | 4 | 1172 | |||
NM_001243752.1 | NP_001230681.1 | 4 | 1246 | |||
NM_001243753.1 | NP_001230682.1 | 4 | 1050 | |||
NM_001243754.1 | NP_001230683.1 | 4 | 952 | |||
NM_001243755.1 | NP_001230684.1 | 4 | 868 | |||
NM_001305629.1 | NP_001292558.1 | 3 | 1096 | |||
NM_024033.3 | NP_076938.2 | 4 | 1135 | |||
XM_017012589.1 | XP_016868078.1 | 4 | 1258 | |||
XM_017012590.1 | XP_016868079.1 | 4 | 1184 | |||
XM_017012591.1 | XP_016868080.1 | 4 | 1038 | |||
XM_017012592.1 | XP_016868081.1 | 3 | 1219 | |||
XM_017012593.1 | XP_016868082.1 | 4 | 1088 | |||
XM_017012594.1 | XP_016868083.1 | 4 | 1295 | |||
GenBank mRNA | AK026103.1 | - | 4 | 1261 | ||
AK130795.1 | - | 2 | 961 | |||
BC000168.2 | - | 4 | 1004 | 2900206 | ||
BC067350.1 | - | 4 | 1041 | 6568468 |