Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243749.1 | NP_001230678.1 | 3 | 779 | ||
NM_001243751.1 | NP_001230680.1 | 3 | 526 | |||
NM_001243752.1 | NP_001230681.1 | 3 | 600 | |||
NM_001243753.1 | NP_001230682.1 | 3 | 404 | |||
NM_001243754.1 | NP_001230683.1 | 3 | 306 | |||
NM_001243755.1 | NP_001230684.1 | 3 | 222 | |||
NM_024033.3 | NP_076938.2 | 3 | 489 | |||
XM_017012589.1 | XP_016868078.1 | 3 | 612 | |||
XM_017012590.1 | XP_016868079.1 | 3 | 538 | |||
XM_017012591.1 | XP_016868080.1 | 3 | 392 | |||
XM_017012593.1 | XP_016868082.1 | 3 | 442 | |||
XM_017012594.1 | XP_016868083.1 | 3 | 649 | |||
XM_017012595.1 | XP_016868084.1 | 3 | 521 | |||
GenBank mRNA | AK026103.1 | - | 3 | 615 | ||
AK130795.1 | - | 1 | 315 | |||
AL708733.1 | - | 3 | 222 | |||
BC000168.2 | - | 3 | 358 | 2900206 | ||
BC067350.1 | - | 3 | 395 | 6568468 | ||
BG770658.1 | - | 3 | 389 | |||
BM727569.1 | - | 2 | 320 | |||
BP213903.1 | - | 3 | 387 | |||
BQ423284.1 | - | 239 | ||||
CB961457.1 | - | 3 | 516 | |||
DA213716.1 | - | 3 | 483 | |||
DB129229.1 | - | 3 | 480 |