Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256737.1 | NP_001243666.1 | 4 | 711 | ||
NM_001256738.1 | NP_001243667.1 | 4 | 501 | |||
NM_001256739.1 | NP_001243668.1 | 3 | 480 | |||
NM_024034.4 | NP_076939.3 | 4 | 654 | |||
NR_046353.1 | 2 | 287 | ||||
GenBank mRNA | AF123764.1 | - | 4 | 553 | ||
AF123765.1 | - | 4 | 596 | |||
AF277566.1 | - | 4 | 609 | |||
AK056251.1 | - | 4 | 644 | |||
AK225166.1 | - | 4 | 644 | |||
AK293484.1 | - | 3 | 430 | |||
AK297348.1 | - | 2 | 189 | |||
AK299259.1 | - | 5 | 566 | |||
AK299700.1 | - | 4 | 660 | |||
BC000199.1 | - | 4 | 630 | 3352559 | ||
BC009014.1 | - | 4 | 593 | 4155653 |