Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311311.1 | NP_001298240.1 | 4 | 988 | ||
NM_024006.5 | NP_076869.1 | 3 | 904 | |||
NM_206824.2 | NP_996560.1 | 2 | 794 | |||
GenBank mRNA | AF176924.1 | - | 4 | 928 | ||
AK129513.1 | - | 2 | 592 | |||
AY466113.1 | - | 3 | 846 | |||
BC000828.1 | - | 3 | 634 | |||
BC002911.2 | - | 3 | 672 | 3944530 |