Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311311.1 | NP_001298240.1 | 4 | 872 | ||
NM_024006.5 | NP_076869.1 | 3 | 788 | |||
NM_206824.2 | NP_996560.1 | 2 | 678 | |||
GenBank mRNA | AF176924.1 | - | 4 | 812 | ||
AK129513.1 | - | 2 | 476 | |||
AK289790.1 | - | 2 | 577 | |||
AY466113.1 | - | 3 | 730 | |||
BC000828.1 | - | 3 | 518 | |||
BC002911.2 | - | 3 | 556 | 3944530 | ||
BI822140.1 | - | 3 | 745 | |||
CD249837.1 | - | 479 | ||||
CN484751.1 | - | 4 | 599 |