Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258217.1 | NP_001245146.1 | 3 | 1134 | ||
NM_001258218.1 | NP_001245147.1 | 2 | 740 | |||
NM_001258219.1 | NP_001245148.1 | 3 | 870 | |||
NM_001258220.1 | NP_001245149.1 | 2 | 476 | |||
NM_024039.2 | NP_076944.1 | 3 | 976 | |||
XM_005256797.2 | XP_005256854.1 | 3 | 1298 | |||
XM_017025033.1 | XP_016880522.1 | 2 | 2154 | |||
XM_017025034.1 | XP_016880523.1 | 3 | 1653 | |||
XM_017025035.1 | XP_016880524.1 | 4 | 1144 | |||
GenBank mRNA | AK056072.1 | - | 2 | 718 | ||
AK289939.1 | - | 3 | 976 | |||
AL136906.1 | - | 3 | 942 | |||
BC000229.1 | - | 2 | 526 | 3351245 | ||
CR533460.1 | - | 1 | 389 |