Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042610.2 | NP_001036075.1 | 4 | 1402 | ||
NM_001288708.1 | NP_001275637.1 | 5 | 1674 | |||
NM_001288709.1 | NP_001275638.1 | 4 | 1561 | |||
NM_024043.3 | NP_076948.2 | 4 | 1322 | |||
GenBank mRNA | AK022644.1 | - | 4 | 1228 | ||
AK090965.1 | - | 3 | 1941 | |||
AK096507.1 | - | 1 | 2138 | |||
AL713678.1 | - | 1 | 827 | |||
BC000700.2 | - | 4 | 1322 | 3350198 |