Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199815.1 | NP_001186744.1 | 4 | 645 | ||
NM_001199816.1 | NP_001186745.1 | 3 | 489 | |||
NM_001199817.1 | NP_001186746.1 | 2 | 343 | |||
NM_024051.3 | NP_076956.1 | 4 | 625 | |||
NR_037669.1 | 3 | 479 | ||||
GenBank mRNA | AK021779.1 | - | 3 | 1104 | ||
AK315608.1 | - | 4 | 616 | |||
BC000625.2 | - | 4 | 620 | 3346822 | ||
BC005356.1 | - | 3 | 477 | 4102029 | ||
BC013937.1 | - | 4 | 586 | 3997629 | ||
BC019243.1 | - | 4 | 617 | 3828006 | ||
BE407390.1 | - | 4 | 574 | |||
BE885397.1 | - | 2 | 303 | |||
BG527593.1 | - | 3 | 468 |