Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA5L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024063.2 | NP_076968.2 | 6 | 2031 | ||
NR_027635.1 | 6 | 2061 | ||||
NR_136645.1 | 6 | 2038 | ||||
NR_136646.1 | 7 | 2256 | ||||
NR_136647.1 | 7 | 2223 | ||||
NR_136648.1 | 7 | 2226 | ||||
GenBank mRNA | AF086107.1 | - | 1 | 72 | ||
AK023232.1 | - | 6 | 2007 | |||
AK225463.1 | - | 6 | 1977 | |||
BC000981.2 | - | 6 | 1964 | 2985725 | ||
BC051861.2 | - | 6 | 1976 |