Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECISBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282688.1 | NP_001269617.1 | 7 | 1150 | ||
NM_001282689.1 | NP_001269618.1 | 7 | 1032 | |||
NM_001282690.1 | NP_001269619.1 | 7 | 1230 | |||
NM_024077.4 | NP_076982.3 | 7 | 1153 | |||
XM_005252196.2 | XP_005252253.1 | 7 | 1003 | |||
XM_005252202.3 | XP_005252259.1 | 3 | 385 | |||
XM_006717282.2 | XP_006717345.1 | 7 | 1086 | |||
XM_011519000.2 | XP_011517302.1 | 7 | 1207 | |||
XM_011519001.1 | XP_011517303.1 | 7 | 1207 | |||
XM_011519002.1 | XP_011517304.1 | 7 | 1093 | |||
XM_011519003.1 | XP_011517305.1 | 7 | 1090 | |||
XM_017015122.1 | XP_016870611.1 | 7 | 1120 | |||
XM_017015123.1 | XP_016870612.1 | 7 | 1006 | |||
XM_017015124.1 | XP_016870613.1 | 7 | 1003 | |||
XM_017015125.1 | XP_016870614.1 | 5 | 708 | |||
GenBank mRNA | AB208940.1 | - | 7 | 1073 | ||
AF380995.1 | - | 7 | 1106 | |||
AK090608.1 | - | 7 | 1120 | |||
AK290182.1 | - | 7 | 999 | |||
AK299161.1 | - | 7 | 1241 | |||
AK302852.1 | - | 7 | 1148 | |||
AL136881.1 | - | 7 | 986 | |||
BC001189.1 | - | 7 | 869 | |||
BC036109.1 | - | 7 | 1115 | 5296292 | ||
BC068466.1 | - | 7 | 1088 | |||
BC099900.1 | - | 7 | 972 |