Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057093.1 | NP_476434.1 | 3 | 603 | ||
NM_057094.1 | NP_476435.1 | 4 | 409 | |||
GenBank mRNA | AF166331.1 | - | 4 | 403 | ||
BC006285.2 | - | 3 | 603 | 3960073 | ||
BF726935.1 | - | 4 | 417 | |||
BT007447.1 | - | 3 | 374 |