Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077198.2 | NP_001070666.1 | 5 | 415 | ||
NM_024085.4 | NP_076990.4 | 4 | 363 | |||
NR_104255.1 | 5 | 415 | ||||
GenBank mRNA | AK056595.1 | - | 5 | 356 | ||
AK289755.1 | - | 5 | 310 | |||
AK302521.1 | - | 4 | 254 | |||
BC065534.1 | - | 5 | 353 | 5787445 | ||
BX537984.1 | - | 4 | 284 | |||
BX538192.1 | - | 4 | 258 | |||
BX538198.1 | - | 5 | 192 | |||
DC347583.1 | - | 4 | 363 |