Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077198.2 | NP_001070666.1 | 8 | 837 | ||
NM_024085.4 | NP_076990.4 | 7 | 785 | |||
NR_104255.1 | 8 | 837 | ||||
GenBank mRNA | AK025822.1 | - | 2 | 139 | ||
AK056595.1 | - | 8 | 778 | |||
AK289755.1 | - | 8 | 732 | |||
AK302521.1 | - | 7 | 676 | |||
AK315553.1 | - | 2 | 149 | |||
AL833865.1 | - | 1 | 34 | |||
BC065534.1 | - | 8 | 775 | 5787445 | ||
BP284912.1 | - | 4 | 495 | |||
BX537984.1 | - | 7 | 706 | |||
BX538192.1 | - | 7 | 680 | |||
BX538198.1 | - | 8 | 614 |