Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077198.2 | NP_001070666.1 | 13 | 2341 | ||
NM_024085.4 | NP_076990.4 | 12 | 2289 | |||
NR_104255.1 | 13 | 2282 | ||||
GenBank mRNA | AK021732.1 | - | 3 | 427 | ||
AK025822.1 | - | 7 | 1643 | |||
AK027448.1 | - | 2 | 266 | |||
AK056595.1 | - | 13 | 2282 | |||
AK289755.1 | - | 13 | 2236 | |||
AK302521.1 | - | 12 | 2180 | |||
AL833865.1 | - | 6 | 1538 | |||
BC001098.2 | - | 5 | 816 | |||
BC021202.2 | - | 4 | 647 | |||
BC065534.1 | - | 13 | 2220 | 5787445 | ||
BX537984.1 | - | 12 | 2210 | |||
BX538192.1 | - | 12 | 2184 | |||
BX538198.1 | - | 13 | 2118 |