Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080432.2 | NP_001073901.1 | 3 | 518 | ||
XM_011523313.2 | XP_011521615.1 | 3 | 482 | |||
XM_011523314.2 | XP_011521616.1 | 3 | 482 | |||
XM_011523315.2 | XP_011521617.1 | 3 | 482 | |||
XM_011523316.2 | XP_011521618.1 | 3 | 482 | |||
XM_017023654.1 | XP_016879143.1 | 3 | 482 | |||
XM_017023655.1 | XP_016879144.1 | 3 | 482 | |||
XM_017023656.1 | XP_016879145.1 | 3 | 482 | |||
XM_017023657.1 | XP_016879146.1 | 3 | 482 | |||
XM_017023658.1 | XP_016879147.1 | 3 | 482 | |||
GenBank mRNA | AB051539.1 | - | 3 | 302 | ||
AK096554.1 | - | 3 | 325 |