Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024096.1 | NP_077001.1 | 2 | 217 | ||
NR_134470.1 | 2 | 242 | ||||
NR_134471.1 | 2 | 256 | ||||
GenBank mRNA | AF210430.1 | - | 2 | 190 | ||
AF212242.1 | - | 2 | 188 | |||
AK024843.1 | - | 2 | 185 | |||
AY453409.1 | - | 2 | 217 | |||
BC001344.1 | - | 2 | 217 | 3463032 | ||
BE798090.1 | - | 269 | ||||
BE897551.1 | - | 2 | 182 | |||
CR457335.1 | - | 2 | 145 |