Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001887.3 | NP_001878.1 | 4, 5 | 500 | ||
XM_011529899.2 | XP_011528201.1 | 4, 5 | 566 | |||
GenBank mRNA | BC036790.1 | - | 4, 5 | 547 | 5752689 | |
CR456425.1 | - | 4, 5 | 469 | |||
U35340.1 | - | 4, 5 | 502 |